Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p53 Monoclonal Antibody (PAb 240), Invitrogen™

Catalog No. PIMA515244
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515244 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515244 Supplier Thermo Scientific Supplier No. MA515244

Mouse Monoclonal Antibody

MA5-15244 targets p53 in FACS, IHC (P), IP, and WB applications and shows reactivity with Human samples. The MA5-15244 immunogen is mouse p53 protein corresponding to aa14-289.

p53 Monoclonal antibody specifically detects p53 in Human, Mouse, Rhesus Monkey samples. It is validated for ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p53
Applications ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone PAb 240
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene Tp53
Gene Accession No. P02340, P04637, P56424
Gene Alias Antigen NY-CO-13; bbl; BCC7; bfy; bhy; cellular tumor antigen p53; Cys 51 Stop; EGK_08142; FLJ92943; HGNC11998; I79_002739; LFS1; Li-Fraumeni syndrome; mutant p53; mutant tumor protein 53; OTTMUSP00000006194; p44; p53; p53 cellular tumor antigen; p53 protein; p53 tumor suppressor; p53 tumor suppressor phosphoprotein; phosphoprotein p53; Tp53; tp53.L; transformation related protein 53; transformation-related protein 53; Trp248; Trp53; tumor protein 53; tumor protein p53; tumor protein p53 (Li-Fraumeni syndrome); tumor protein p53 L homeolog; tumor suppressor p53; tumor suppressor p53 phosphoprotein; tumor suppressor protein p53; tumor supressor p53; Tumour Protein p53; XELAEV_180196761mg; Xp53; Xrel3
Gene Symbols Tp53, Trp53
Host Species Mouse
Immunogen Murine p53 protein corresponding to aa14-289
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22059, 7157, 716170
Target Species Human, Mouse, Rhesus Monkey
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.