Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ p53 (Pantropic), Mouse, Unlabeled, Clone: DO-1,

Catalog No. OP4320UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OP4320UG 20 μg
OP43100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. OP4320UG Supplier MilliporeSigma™ Supplier No. OP4320UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recognizes the ∽53kDa wild-type and mutant p53 protein in A431 cells and breast carcinoma tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p53 (Pantropic)
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone DO-1
Concentration Please refer to vial label for lot-specific concentration
Conjugate Unconjugated
Formulation In 0.05M sodium phosphate buffer, 0.2% gelatin, ≤0.1% Sodium Azide.
Host Species Mouse
Immunogen wild type, recombinant, human p53
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Feline
Content And Storage 2°-8°C
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.