Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p53 Rat anti-Mouse, Unconjugated, Clone: POE316A/E9, MilliporeSigma™

Catalog No. MABE18081MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE18081MI 100 μg
MABE18082MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE18081MI Supplier MilliporeSigma Supplier No. MABE1808100UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

p53 Monoclonal antibody specifically detects p53 in Mouse samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen p53
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone POE316A/E9
Conjugate Unconjugated
Gene Accession No. NP_035770.2
Gene Alias Cellular tumor antigen p53;Tumor suppressor p53
Gene Symbols Tp53, P53, Trp53
Host Species Rat
Immunogen GST/His-tagged full-length recombinant mouse p53 protein.
Molecular Weight of Antigen 43.46 kDa calculated.
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone POE316A/E9 is a rat monoclonal antibody that detects murine p53.
Target Species Mouse
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.