Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ p53DINP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103262
Description
Antibody detects endogenous levels of total TP53INP1.
A novel p53 inducible gene was identified and designated p53DINP1 (for p53-dependent damage-inducible nuclear protein). p53DINP1 may regulate p53-dependent apoptosis through phosphorylation at Ser46 and induction of p53AIP1. p53DINP1/SIP is expressed in many tissues and induced by a variety of stress agents including UV stress, mutagenic stress, heat shock, and oxidative stress.
Specifications
p53DINP1 | |
Polyclonal | |
Unconjugated | |
TP53INP1 | |
2700057G22Rik; p53-dependent damage-inducible nuclear protein 1; p53DINP1; p53-inducible p53DINP1; Sip; SIP18; SIP27; Stinp; stress induced protein; stress-induced protein; TEAP; thymus expressed acidic protein; thymus-expressed acidic protein; TP53DINP1; TP53INP1; TP53INP1A; TP53INP1B; transformation related protein 53 inducible nuclear protein 1; Trp53inp1; tumor protein p53 inducible nuclear protein 1; tumor protein p53-inducible nuclear protein 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
60599, 94241 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96A56, Q9QXE4 | |
TP53INP1, Trp53inp1 | |
A synthesized peptide derived from human TP53INP1(Accession Q96A56), corresponding to amino acid residues G166-R216. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction