Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ p53DINP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582221
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582221 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582221 Supplier Invitrogen™ Supplier No. PA582221
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VEAQNEMGQH IHCYVAALAA HTTFLEQPKS FRPSQWIKEH SERQPLNRNS LRRQNLTRDC HPRQVKHNGW VVHQPC Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 82%.

A novel p53 inducible gene was identified and designated p53DINP1 (for p53-dependent damage-inducible nuclear protein). p53DINP1 may regulate p53-dependent apoptosis through phosphorylation at Ser46 and induction of p53AIP1. p53DINP1/SIP is expressed in many tissues and induced by a variety of stress agents including UV stress, mutagenic stress, heat shock, and oxidative stress.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p53DINP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TP53INP1
Gene Accession No. Q96A56
Gene Alias 2700057G22Rik; p53-dependent damage-inducible nuclear protein 1; p53DINP1; p53-inducible p53DINP1; Sip; SIP18; SIP27; Stinp; stress induced protein; stress-induced protein; TEAP; thymus expressed acidic protein; thymus-expressed acidic protein; TP53DINP1; TP53INP1; TP53INP1A; TP53INP1B; transformation related protein 53 inducible nuclear protein 1; Trp53inp1; tumor protein p53 inducible nuclear protein 1; tumor protein p53-inducible nuclear protein 1
Gene Symbols TP53INP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TP53INP1. Recombinant protein control fragment (Product #RP-88834).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94241
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.