Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ p53DINP1 Recombinant Rabbit Monoclonal Antibody (JG37-64)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA534751
Description
Positive Control: SH-SY-5Y, HUVEC, HepG2, Mouse stomach tissue, Human stomach tissue, human liver cancer tissue, human kidney tissue, human pancreas tissue.
A novel p53 inducible gene was identified and designated p53DINP1 (for p53-dependent damage-inducible nuclear protein). p53DINP1 may regulate p53-dependent apoptosis through phosphorylation at Ser46 and induction of p53AIP1. p53DINP1/SIP is expressed in many tissues and induced by a variety of stress agents including UV stress, mutagenic stress, heat shock, and oxidative stress.
Specifications
p53DINP1 | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q96A56 | |
TP53INP1 | |
Recombinant protein within Human p53 DINP1 aa 80-188. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot, Immunocytochemistry | |
JG37-64 | |
Unconjugated | |
TP53INP1 | |
2700057G22Rik; p53-dependent damage-inducible nuclear protein 1; p53DINP1; p53-inducible p53DINP1; Sip; SIP18; SIP27; Stinp; stress induced protein; stress-induced protein; TEAP; thymus expressed acidic protein; thymus-expressed acidic protein; TP53DINP1; TP53INP1; TP53INP1A; TP53INP1B; transformation related protein 53 inducible nuclear protein 1; Trp53inp1; tumor protein p53 inducible nuclear protein 1; tumor protein p53-inducible nuclear protein 1 | |
Rabbit | |
Protein A | |
RUO | |
94241 | |
-20°C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction