Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p53R2 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300079APC
Description
p53R2 Polyclonal antibody specifically detects p53R2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)Specifications
p53R2 | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry-Paraffin | |
EC 1.17.4.1, MGC102856, MGC42116, MTDPS8A, MTDPS8B, p53-inducible ribonucleotide reductase small subunit 2 homolog, p53-inducible ribonucleotide reductase small subunit 2-like protein, p53R2, P53R2DKFZp686M05248, PEOA5, ribonucleoside-diphosphate reductase subunit M2 B, ribonucleotide reductase M2 B (TP53 inducible), TP53-inducible ribonucleotide reductase M2 B | |
Produced in rabbits immunized with purified, recombinant Human p53R2 (Uniprot#: Q7LG56-1; Met 1-Phe 351) | |
0.1 mL | |
Apoptosis, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Hypoxia, Modulation of DNA Pools | |
50484 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction