Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p57 Kip2 Antibody (KIP2/8169R), Novus Biologicals™

Rabbit Monoclonal Antibody
$247.50 - $474.50
Specifications
Antigen | p57 Kip2 |
---|---|
Clone | KIP2/8169R |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 0.25-0.5 μg/mL |
Applications | Immunohistochemistry (Paraffin) |
Description
p57 Kip2 Monoclonal antibody specifically detects p57 Kip2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
p57 Kip2 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
10mM PBS with 0.05% BSA | |
1028 | |
IgG κ | |
Protein A or G purified |
KIP2/8169R | |
Immunohistochemistry-Paraffin 0.25-0.5 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Beckwith-Wiedemann syndrome, BWS, cyclin-dependent kinase inhibitor 1C, cyclin-dependent kinase inhibitor 1C (p57, Kip2), Cyclin-dependent kinase inhibitor p57, KIP2BWCR, p57, p57Kip2, WBS | |
Recombinant full-length human p57 Kip2 protein | |
Primary | |
Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title