Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p57Kip2 Mouse anti-Human, Mouse, Rat, Clone: 1A12H3, Proteintech
SDP

Catalog No. 501737194 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7194 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7194 Supplier Proteintech Group Inc Supplier No. 667941IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene is part of a gene cluster on chromosome Xp11.23. The encoded protein contains a zinc finger motif often found in transcriptional regulators, however, its exact function is not known. Alternative splicing results in multiple transcript variants encoding the same protein.

Specifications

Antigen p57Kip2
Applications Western Blot
Classification Monoclonal
Clone 1A12H3
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CDKN1C
Gene Accession No. P49918, P49919
Gene Alias BWCR, BWS, CDKN1C, KIP2, p57, p57 Kip2, p57Kip2, WBS
Gene Symbols CDKN1C
Host Species Mouse
Immunogen p57Kip2 Fusion Protein Ag19927
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1028, 12577, 246060
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.