Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p57kip2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501562374
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-2374 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-2374 Supplier Bethyl Laboratories, Inc Supplier No. A302346AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The cyclin-dependent kinase inhibitor 1C (CDKN1C), also known as p57KIP2, is a tight-binding, strong inhibitor of several G1 cyclin/CDK complexes and a negative regulator of cell proliferation. Mutations in this gene are implicated in sporadic cancers and Beckwith-Wiedemann syndrome, a cancer-predisposing syndrome, suggesting that this gene is a tumor suppressor candidate. CDKN1C is widely expressed during organogenesis with its expression declining after maturity. It is part of the CIP/KIP family of cyclin-dependent inhibitors (CKIs) which also includes CDKN1A and CDKN1B.

Specifications

Antigen p57kip2
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene CDKN1C
Gene Accession No. P49918
Gene Alias p57 Kip2
Gene Symbols CDKN1C
Host Species Rabbit
Immunogen Between 225 and 275
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1028
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.