Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p57Kip2Ab-6 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1062P1ABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1062P1ABX 100 μL Concentrated 1mg/mL
MS1062P0 100 μL Concentrated 200μg/mL
MS1062PABX 200 μL Concentrated 1mg/mL
MS1062P1 500 μL Concentrated 200μg/mL
MS1062P 1 mL Concentrated 200μg/mL
MS1062R7 7 mL Prediluted -
MS1062R1 1 mL Prediluted -
MS1062RQ 12 mL Ready-to-Use -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS1062P1ABX Supplier Epredia Supplier No. MS1062P1ABX

Deliver accurate, reproducible results with the Epredia™ p57Kip2 Ab-6, Mouse Monoclonal Antibody.

p57Kip2 (or CDKN1C) is a potent tight-binding inhibitor of several G1 cyclin complexes, and is a negative regulator of cell proliferation. The gene encoding human p57Kip2 is located on chromosome 11p15.5, a region implicated in both sporadic cancers, Wilm's tumor, and Beckwith-Wiedemann syndrome (BWS), a cancer syndrome, making it a tumor suppressor candidate. BWS is characterized by numerous growth abnormalities and an increased risk of childhood tumors. Several types of childhood tumors including Wilms' tumor, adrenocortical carcinoma and rhabdomyosarcoma display a specific loss of maternal 11p15 alleles, suggesting that genomic imprinting plays an important part. This region also contains two other imprinted genes, insulin-like growth factor II (IGF-II) and H19, both of which seem to be implicated in adrenal neoplasms.

Host Species: Mouse

Clone: 57P06; same as KP10

Isotype: IgG2b/κ

Species Reactivity: Human and Mouse. Others not known.

Epitope: Not determined

Immunogen: Recombinant human p57Kip2 protein

Molecular Weight: 57kDa

Positive Control: LS174T cells, Colon CA or placenta

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not suitable)
  • Immunoprecipitation (Native verified)

Specifications

Antigen p57superscriptKip2superscript Ab-6
Applications Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone 57P06 ;same as KP10
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant human p57superscriptKip2superscript protein
Quantity 100 μL
Regulatory Status IVD
Research Discipline Cell Cycle
Primary or Secondary Primary
Target Species Human, Mouse
Form Concentrated
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.