Learn More
p600 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc IHC00640

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Horse, Pig, Orangutan, Monkey, Gorilla, Chimpanzee, Northern white-cheeked gibbon and
E3 ubiquitin-protein ligase which is a component of the N-end rule pathway. Recognizes and binds to proteins bearing specific N-terminal residues that are destabilizing according to the N-end rule, leading to their ubiquitination and subsequent degradation. Together with clathrin, forms meshwork structures involved in membrane morphogenesis and cytoskeletal organization. Regulates integrin-mediated signaling. May play a role in activation of FAK in response to cell-matrix interactions. Mediates ubiquitination of ACLY, leading to its subsequent degradation.Specifications
p600 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q5T4S7 | |
UBR4 | |
Between 5133 and 5183 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
UBR4 | |
600 kDa retinoblastoma protein-associated factor, KIAA0462, KIAA1307, N-recognin-4, p600, RBAF600, retinoblastoma-associated factor 600-like protein, retinoblastoma-associated factor of 600 kDa, zinc finger UBR1-type protein 1, zinc finger, UBR1 type 1, ZUBR1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23352 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.