Learn More
p600 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302278A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
E3 ubiquitin-protein ligase which is a component of the N-end rule pathway. Recognizes and binds to proteins bearing specific N-terminal residues that are destabilizing according to the N-end rule, leading to their ubiquitination and subsequent degradation. Together with clathrin, forms meshwork structures involved in membrane morphogenesis and cytoskeletal organization. Regulates integrin-mediated signaling. May play a role in activation of FAK in response to cell-matrix interactions. Mediates ubiquitination of ACLY, leading to its subsequent degradation.Specifications
p600 | |
Polyclonal | |
Unconjugated | |
UBR4 | |
600 kDa retinoblastoma protein-associated factor, KIAA0462, KIAA1307, N-recognin-4, p600, RBAF600, retinoblastoma-associated factor 600-like protein, retinoblastoma-associated factor of 600 kDa, zinc finger UBR1-type protein 1, zinc finger, UBR1 type 1, ZUBR1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23352, 69116 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
A2AN08, Q5T4S7 | |
Ubr4 | |
Between 1350 and 1400 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.