Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p62/SQSTM1 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149954B
Description
p62/SQSTM1 Polyclonal specifically detects p62/SQSTM1 in Human, Mouse, Canine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
p62/SQSTM1 | |
Polyclonal | |
Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin | |
A170, Autophagy Receptor P62, DMRV, EBI3-associated protein of 60 kDa, EBI3-associated protein p60, EBIAP, FTDALS3, NADGP, ORCA, OSIL, oxidative stress induced like, p60PDB3, p62, p62B, Paget disease of bone 3, PDB3, phosphotyrosine independent ligand for the Lck SH2 domain p62, Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa, sequestosome 1, sequestosome-1, Ubiquitin-binding protein p62, ZIP3 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Biotin | |
PBS with 0.05% Sodium Azide | |
SQSTM1 | |
The immunogen for this product maps to a region between residue 325 and 375 of human p62/SQSTM1 using the numbering given in entry NP_003891.1 (GeneID 8878). | |
0.1 mL | |
Adaptive Immunity, Cancer, Chromatin Modifiers, Chromatin Research, DNA Methyltransferases, Epigenetics, Innate Immunity, Tumor Suppressors | |
Human, Mouse, Canine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction