Learn More
P62/SQSTM1 Mouse anti-Human, Clone: 1H5C1, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | P62/SQSTM1 |
---|---|
Clone | 1H5C1 |
Concentration | 1.33 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6618
![]() ![]() |
Proteintech Group Inc
661841IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The molecular weight of p62 is predicted as 48/ 38 kDa, while western blot analyses using this antibody demonstrate the major band around 60-62 kDa in various tissues.
This gene encodes a multifunctional protein that binds ubiquitin and regulates activation of the nuclear factor kappa-B signaling pathway. The protein functions as a scaffolding/adaptor protein in concert with TNF receptor-associated factor 6 to mediate activation of NF-kB in response to upstream signals. Alternatively spliced transcript variants encoding either the same or different isoforms have been identified for this gene. Mutations in this gene result in sporadic and familial Paget disease of bone.Specifications
P62/SQSTM1 | |
1.33 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A170, EBIAP, ORCA, OSIL, p60, P62, P62;SQSTM1, p62B, PDB3, sequestosome 1, SQSTM1, Ubiquitin binding protein p62, ZIP3 | |
SQSTM1 | |
IgG2a | |
Protein A | |
Antibody |
1H5C1 | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q13501 | |
8878 | |
P62/SQSTM1 Fusion Protein Ag13131 | |
Primary | |
-20°C | |
SQSTM1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.