Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p62/SQSTM1, p Thr269, p Ser272 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP229650
Description
p62/SQSTM1 Polyclonal specifically detects p62/SQSTM1 in Human samples. It is validated for Western Blot.Specifications
p62/SQSTM1 (p Thr269, p Ser272) | |
Polyclonal | |
Western Blot 1:1000 | |
Q13501 | |
SQSTM1 | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding p62/SQSTM1 conjugated to KLH. Accession # Q13501 | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Specific for the ∼62k p62 protein phosphorylated at Thr269 and Ser272. Immunolabeling is blocked by preadsorption with the phosphopeptide used as antigen, but not by the corresponding non-phosphopeptide | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
A170, EBI3-associated protein of 60 kDa, EBI3-associated protein p60, EBIAP, ORCA, OSIL, oxidative stress induced like, p60PDB3, p62, p62B, Paget disease of bone 3, phosphotyrosine independent ligand for the Lck SH2 domain p62, Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa, sequestosome 1, sequestosome-1, Ubiquitin-binding protein p62, ZIP3 | |
Rabbit | |
62 kDa | |
0.1 mL | |
Autophagy, Signal Transduction | |
8878 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction