Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p66 alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$405.00 - $670.00
Specifications
Antigen | p66 alpha |
---|---|
Concentration | 0.1mg/mL |
Applications | ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
p66 alpha Polyclonal specifically detects p66 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP).Specifications
p66 alpha | |
ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
FLJ20085, FLJ21017, GATA zinc finger domain containing 2A, GATA zinc finger domain-containing protein 2A, Hp66alpha, p66 alpha, p66alpha, transcriptional repressor p66-alpha | |
GATAD2A | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
0.1mg/mL | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat | |
Q86YP4 | |
54815 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:RRKLAFRSGEARDWSNGAVLQASSQLSRGSATTPRGVLHTFSPSPKLQNSASATALVSRTGRHSERTVSAGKGSATSNWKKTPLSTGGTLAFVSPSLAVHKSSSAVDRQREYLLDMIPPRSIP | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
68 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title