Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p66 beta Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen p66 beta
Concentration 0.5 mg/mL
Dilution Western Blot 2-4 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056387T
SDP
View Documents
Novus Biologicals
NB10056387SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NB10056387
SDP
View Documents
Novus Biologicals
NB10056387
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

p66 beta Polyclonal specifically detects p66 beta in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

p66 beta
Western Blot 2-4 ug/ml
Polyclonal
Purified
RUO
PBS containing 0.05% BSA with 0.05% Sodium Azide
57459
This antibody was developed against a synthetic peptide corresponding to aa 445-460 of mouse p66b.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Human, Mouse
FLJ37346, GATA zinc finger domain containing 2B, GATA zinc finger domain-containing protein 2B, KIAA1150, MGC138257, MGC138285, p66/p68, P66beta, RP11-216N14.6, transcription repressor p66 beta component of the MeCP1 complex, transcriptional repressor p66-beta
GATAD2B
IgG
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.