Learn More
p66beta/GATAD2B Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301281A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Methylation of cytosine at CpG dinucleotides is a common feature of many higher eukaryotic genomes (1-3). Increasing evidence indicates that recruitement of histone deacetylase complexes by methyl-CpG-binding proteins (MeCP) is a major mechanism of methylated DNA silencing. P66, a zinc finger-containing protein is one of the components of the MeCP1 complex (4). P66 and p68, the two components of MeCP1 are different forms of the same zinc finger-containing protein and are conserved from C.elegans to humans. Homologs of p66 from different organisms revealed two highly conserved regions, CR1 and CR2. While CR1 is involved in the association of p66 with other MeCP1 components, CR2 plays an important role in targeting p66 and MBD3 to specific loci (4).Specifications
p66beta/GATAD2B | |
Polyclonal | |
Unconjugated | |
GATAD2B | |
AL118180, C430014D17Rik, KIAA1150, P66beta, mKIAA1150 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
229542, 57459 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8VHR5, Q8WXI9 | |
GATAD2B | |
Between 1 and 50 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.