Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P70 S6 Kinase beta Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502208933
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8933 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8933 Supplier Fabgennix Inc Supplier No. S6K101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

P70 S6 Kinase beta Polyclonal antibody specifically detects P70 S6 Kinase beta in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen P70 S6 Kinase beta
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Rps6kb1
Gene Accession No. P23443, P67999, Q8BSK8
Gene Alias 2610318I15Rik; 4732464A07Rik; 70 kDa ribosomal protein S6 kinase 1; 70 kDa ribosomal protein S6 kinase 2; 70kDa; AA959758; AI256796; AI314060; KLS; p70 ribosomal S6 kinase alpha; p70 ribosomal S6 kinase beta; p70 S6 kinase alpha; p70 S6 kinase beta; p70 S6 kinase, alpha; p70 S6KA; p70 S6K-alpha; p70 S6KB; p70 S6K-beta; p70(S6K)-alpha; p70(S6K)-beta; p70/85s6k; p70-alpha; P70-beta; P70-beta-1; P70-beta-2; P70S6K; p70-S6K; p70-S6K 1; p70-S6K 2; P70S6K1; P70S6K2; p70S6Kb; PS6K; ribosomal protein S6 kinase B1; ribosomal protein S6 kinase B2; ribosomal protein S6 kinase beta-1; Ribosomal protein S6 kinase beta-2; ribosomal protein S6 kinase I; ribosomal protein S6 kinase, 70kD, polypeptide 1; ribosomal protein S6 kinase, 70kD, polypeptide 2; ribosomal protein S6 kinase, 70kDa, polypeptide 1; ribosomal protein S6 kinase, 70kDa, polypeptide 2; ribosomal protein S6 kinase, polypeptide 1; ribosomal protein S6 kinase, polypeptide 2; RPS6KB1; Rps6kb2; S6 kinase; S6 kinase 2; S6 kinase-related kinase; S6K; S6K1; S6K2; S6K-beta; S6K-beta-1; S6K-beta2; S6K-beta-2; serine/threonine kinase 14 alpha; serine/threonine-protein kinase 14 beta; serine/threonine-protein kinase 14A; Serine/threonine-protein kinase 14B; SRK; STK14A; STK14B
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 400-450 on human Ribosomal protein S6 kinase beta protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6198, 72508, 83840
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.