Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p70 S6 Kinase/S6K Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100293FR
Description
p70 S6 Kinase/S6K Polyclonal antibody specifically detects p70 S6 Kinase/S6K in Human, Mouse samples. It is validated for Western BlotSpecifications
p70 S6 Kinase/S6K | |
Polyclonal | |
Western Blot | |
EC 2.7.11, EC 2.7.11.1, p70 ribosomal S6 kinase alpha, p70 S6 kinase alpha, p70 S6 kinase, alpha 1,70 kDa ribosomal protein S6 kinase 1, p70 S6 kinase, alpha 2, p70 S6KA, p70 S6K-alpha, p70(S6K)-alpha, p70-alpha, p70-S6K, P70S6K1, PS6K, ribosomal protein S6 kinase beta-1, Ribosomal protein S6 kinase I, ribosomal protein S6 kinase, 70kD, polypeptide 1, ribosomal protein S6 kinase, 70kDa, polypeptide 1, S6K, S6K1p70-S6K 1, S6K-beta-1, serine/threonine kinase 14 alpha, Serine/threonine-protein kinase 14A, STK14A | |
The immunogen recognized by this antibody maps to a region between residue 475 and the C-terminus (residue 525) of human Ribosomal Protein S6 Kinase, 70 kDa, polypeptide 1 using the numbering given in entry NP_003152.1 (GeneID 6198). | |
0.1 mL | |
Autophagy, Cancer, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction | |
6198 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction