Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p70 S6 Kinase/S6K, p Ser411 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$268.00 - $426.00
Specifications
Antigen | p70 S6 Kinase/S6K (p Ser411) |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017872
![]() |
Novus Biologicals
NB100821200.025ML |
0.025 mL |
Each for $268.00
|
|
|||||
NB10082120
![]() |
Novus Biologicals
NB10082120 |
0.05 mL |
Each for $426.00
|
|
|||||
Description
p70 S6 Kinase/S6K Polyclonal specifically detects p70 S6 Kinase/S6K in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
p70 S6 Kinase/S6K (p Ser411) | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100 | |
Polyclonal | |
Rabbit | |
Autophagy, mTOR Pathway, Phospho Specific, Protein Kinase | |
P23443 | |
6198 | |
IgG | |
Affinity Purified | |
80 kDa |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Mouse, Human, Rat | |
EC 2.7.11, EC 2.7.11.1, p70 ribosomal S6 kinase alpha, p70 S6 kinase alpha, p70 S6 kinase, alpha 1,70 kDa ribosomal protein S6 kinase 1, p70 S6 kinase, alpha 2, p70 S6KA, p70 S6K-alpha, p70(S6K)-alpha, p70-alpha, p70-S6K, P70S6K1, PS6K, ribosomal protein S6 kinase beta-1, Ribosomal protein S6 kinase I, ribosomal protein S6 kinase, 70kD, polypeptide 1, ribosomal protein S6 kinase, 70kDa, polypeptide 1, S6K, S6K1p70-S6K 1, S6K-beta-1, serine/threonine kinase 14 alpha, Serine/threonine-protein kinase 14A, STK14A | |
RPS6KB1 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title