Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p70S6 Kinase Mouse anti-Human, Unconjugated, Clone: 10H11.1, MilliporeSigma™

Catalog No. MABS168725 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1687-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1687-25 Supplier MilliporeSigma Supplier No. MABS168725UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-p70S6 Kinase, clone 10H11.1, Cat. No. MABS1687, is a mouse monoclonal antibody that detects Ribosomal protein S6 kinase beta-1 and has ben tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p70S6 Kinase
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 10H11.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Ribosomal protein S6 kinase beta-1;EC 2.7.11.1;S6K-beta-1;S6K1;70 kDa ribosomal protein S6 kinase 1;P70S6K1;p70-S6K 1;Ribosomal protein S6 kinase I;Serine/threonine-protein kinase 14A;p70 ribosomal S6 kinase alpha;p70 S6 kinase alpha;p70 S6K-alpha;p70 S6KA
Gene Symbols RPS6KB1;STK14A
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to the first 93 amino acids from the N-terminal region of human Ribosomal protein S6 kinase beta-1.
Molecular Weight of Antigen ∽70 kDa observed; 59.14 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_003152
Test Specificity Clone 10H11.1 is a mouse monoclonal antibody that detects human Ribosomal protein S6 kinase beta-1. It targets an epitope within the first 93 amino acids from the N-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.