Learn More
Promega p70S6K Kinase Enzyme System, 1mg
Catalog No. PRV2742 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV2742 1 mg
1 options
Catalog No. PRV2742 Supplier Promega Supplier No. V2742
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human p70S6K was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. p70S6K is responsible for the phosphorylation of 40S ribosomal protein S6 and is ubiquitously expressed in human adult tissues.
  • Easily Screen and Profile p70S6K Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C