Learn More
Promega p70S6Kb Kinase Enzyme System, 10microg
Catalog No. PRV4030 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4030 10 μg
1 options
Catalog No. PRV4030 Supplier Promega Supplier No. V4030
Recombinant full-length human p70S6Kb was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. p70S6Kb is a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases and is activated by mitogenic stimuli.
  • Easily Screen and Profile p70S6Kb Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C