Learn More
Promega p70S6Kb Kinase Enzyme System, 1mg
Catalog No. PRV6456 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6456 1 mg
1 options
Catalog No. PRV6456 Supplier Promega Supplier No. V6456
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human p70S6Kb was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. p70S6Kb is a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases and is activated by mitogenic stimuli.
  • Easily Screen and Profile p70S6Kb Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C