Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p73 Antibody (5B1288) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 28 publications

$206.50 - $499.50

Specifications

Antigen p73
Clone 5B1288
Concentration 1.0 mg/mL
Dilution Western Blot 1-3 ug/ml, Chromatin Immunoprecipitation 1:20 - 1:1000. Use reported in scientific literature (Accardi et al), Immunohistochemistry 1:100 - 1:200. Use reported in scientific literature (PMID 19816568), Immunocytochemistry/Immunofluorescence 1:10-1:2000. Use reported in scientific literature (Sayan et al), Immunoprecipitation 1:20 - 1:1000. Use reported in scientific literature (Sayan et al), Immunohistochemistry-Paraffin 1:100 - 1:200, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000
Applications Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056674T
SDP
View Documents
Novus Biologicals
NB10056674SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10056674
SDP
View Documents
Novus Biologicals
NB10056674
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

p73 Monoclonal specifically detects p73 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

p73
1.0 mg/mL
Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Apoptosis, Cancer, Tumor Suppressors
O15350
7161
This antibody was raised against full-length human p73. The epitope is thought to lie around the center of the molecule (NP_005418).
Primary
Store at -20C. Avoid freeze-thaw cycles.
73 kDa
5B1288
Western Blot 1-3 ug/ml, Chromatin Immunoprecipitation 1:20 - 1:1000. Use reported in scientific literature (Accardi et al), Immunohistochemistry 1:100 - 1:200. Use reported in scientific literature (PMID 19816568), Immunocytochemistry/Immunofluorescence 1:10-1:2000. Use reported in scientific literature (Sayan et al), Immunoprecipitation 1:20 - 1:1000. Use reported in scientific literature (Sayan et al), Immunohistochemistry-Paraffin 1:100 - 1:200, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000
Monoclonal
Purified
RUO
Human, Mouse
p53-related protein, P73p53-like transcription factor, TA p73, TAp73, tumor protein p73
TP73
IgG1 κ
Protein G purified
It reacts with alpha, beta, gamma and delta isoforms of mouse and human p73 as well as the dominant negative p73 (Costanzo, et al, 2002). Because of its reactivity pattern it may be regarded as anti-pan p73. NB100-56674 has been shown to recognize all the known alternative splicing variants of human and mouse p73 (Costanzo et al, 2002). The antibody does not cross react with p53.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.