Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p73 Antibody (5B429) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 56 publications
$206.50 - $501.50
Specifications
Antigen | p73 |
---|---|
Clone | 5B429 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 2 ug/ml, Simple Western 1:25, Flow Cytometry 1 ug/mL. Use reported in scientific literature (PMID 21152443), Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 5-15 ug/ml, Immunohistochemistry-Frozen 5-15 ug/ml, Flow (Intracellular), Chromatin Immunoprecipitation (ChIP) 1:20-1:1000, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224737SS
![]() |
Novus Biologicals
NBP224737SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP224737
![]() |
Novus Biologicals
NBP224737 |
0.1 mg |
Each for $501.50
|
|
|||||
Description
p73 Monoclonal specifically detects p73 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready.Specifications
p73 | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer, Tumor Suppressors | |
O15350 | |
7161 | |
This antibody was raised against full-length human p73. It reacts with an epitope that is located on the N-terminal region of human p73. It does not cross react with p53. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
5B429 | |
Western Blot 2 ug/ml, Simple Western 1:25, Flow Cytometry 1 ug/mL. Use reported in scientific literature (PMID 21152443), Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 5-15 ug/ml, Immunohistochemistry-Frozen 5-15 ug/ml, Flow (Intracellular), Chromatin Immunoprecipitation (ChIP) 1:20-1:1000, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
p53-related protein, P73p53-like transcription factor, TA p73, TAp73, tumor protein p73 | |
TP73 | |
IgG1 κ | |
Protein G purified | |
p73 antibody (clone 5B429) recognizes all of the TAp73 isoforms i.e. isoforms with an intact NH2-terminal transactivation domain (Thottassery et el, 2006, PMID 16505115). Sayan et al 2005 (PMID 15781249) showed that p73 IMG-246/ NBP2-24737 reacted with all tested TAp73 isoforms but not deltaNp73 whereas p73 antibody clone 5B1288 (NB100-56674/ IMG-259A ) showed reactivity against all p73 isoforms including delta Np73. Caspase-cleaved p73 fragments, including 55 kDa and 45 kDa, have been described in literature (Sayan et al, 2008). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title