Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p73 Antibody (5B429), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP224737MFV450
Description
p73 Monoclonal antibody specifically detects p73 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
p73 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
5B429 | |
Flow Cytometry, Flow (Intracellular) | |
p53-related protein, P73p53-like transcription factor, TA p73, TAp73, tumor protein p73 | |
This antibody was raised against full-length human p73. It reacts with an epitope that is located on the N-terminal region of human p73. It does not cross react with p53. | |
0.1 mL | |
Apoptosis, Cancer, Phospho Specific, Tumor Suppressors | |
7161 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction