Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p73 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100420JF646
Description
p73 Polyclonal antibody specifically detects p73 in Human samples. It is validated for Western BlotSpecifications
p73 | |
Polyclonal | |
Western Blot | |
p53-related protein, P73p53-like transcription factor, TA p73, TAp73, tumor protein p73 | |
A synthetic peptide mapping to a region between residues 1 and 62 of human tumor protein p73 using the numbering given in Swiss-Prot entry O15350 (GeneID 7161). | |
0.1 mL | |
Apoptosis, Cancer, Phospho Specific, Tumor Suppressors | |
7161 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction