Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p73 Antibody (P73/2531), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308760AF647
Description
p73 Monoclonal antibody specifically detects p73 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
p73 | |
Monoclonal | |
Alexa Fluor 647 | |
p53-related protein, P73p53-like transcription factor, TA p73, TAp73, tumor protein p73 | |
Recombinant full-length human p73 protein (Uniprot: O15350) | |
100 μg | |
Apoptosis, Cancer, Tumor Suppressors | |
7161 | |
Store at 4°C in the dark. | |
IgG2b κ |
Immunohistochemistry (Paraffin) | |
P73/2531 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction