Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ p73 Monoclonal Antibody (5B429)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA516167
Description
p73 Monoclonal Antibody for Western Blot, ICC/IF, IHC (F), IHC (P), Flow, IP, ChIP
P73 (TP73) participates in the apoptotic response to DNA damage. Isoforms containing the transactivation domain are pro-apoptotic, isoforms lacking the domain are anti-apoptotic and block the function of p53 and transactivating p73 isoforms. May be a tumor suppressor protein.
Specifications
p73 | |
Monoclonal | |
1.0 mg/mL | |
PBS with 0.05% sodium azide | |
O15350, Q9JJP2 | |
TP73, Trp73 | |
This antibody was raised against full-length human p73. It reacts with an epitope that is located on the N-terminal region of human p73. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 κ |
ChIP Assay, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
5B429 | |
Unconjugated | |
TP73 | |
deltaNp73; OTTHUMP00000003479; OTTHUMP00000003480; p53-like transcription factor; p53-related protein; P73; RP23-254N4.10; TAp73; Tp73; transformation related protein 73; Trp73; Tumor protein p73 | |
Mouse | |
Protein G | |
RUO | |
22062, 362675, 7161 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction