Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p73 Antibody (5B1288), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056674AF532
Description
p73 Monoclonal specifically detects p73 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
p73 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
TP73 | |
This antibody was raised against full-length human p73. The epitope is thought to lie around the center of the molecule (NP_005418). | |
0.1 mL | |
Primary | |
It reacts with alpha, beta, gamma and delta isoforms of mouse and human p73 as well as the dominant negative p73 (Costanzo, et al, 2002). Because of its reactivity pattern it may be regarded as anti-pan p73. NB100-56674 has been shown to recognize all the known alternative splicing variants of human and mouse p73 (Costanzo et al, 2002). The antibody does not cross react with p53. | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
5B1288 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
p53-related protein, P73p53-like transcription factor, TA p73, TAp73, tumor protein p73 | |
Mouse | |
Protein G purified | |
Apoptosis, Cancer, Phospho Specific, Tumor Suppressors | |
7161 | |
Human, Mouse | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction