Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ p75NTR, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABN1655MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN1655MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1655MI Supplier MilliporeSigma™ Supplier No. ABN1655
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Interferon-induced protein with tetratricopeptide repeats 5 (UniProt Q13325; also known as IFIT-5, Interferon-induced 58kDa protein, P58, Retinoic acid- and interferon-inducible 58kDa protein) is encoded by the IFIT5 (also known as ISG58, RI58) gene (Gene ID 24138) in human. IFIT1, IFIT2, IFIT3, and IFIT5 constitute the family of interferon-stimulated genes (ISGs) that are induced in response to virus infection, stimulation by interferons (IFNs) or double-stranded RNA (dsRNA). The protiens encoded by IFIT1 and IFIT2 are called ISG56 (p56) and ISG54 (p54) in both human and murine species, while the murine IFIT3 (ISG49 or p49) is of a different size than its human counterpart (ISG60 or p60) and no known murine IFIT5 analog exists. IFIT family proteins restrict virus replication by various means. IFIT1 is known to disrupt complex formation between eIF3 and HCV RNA and inhibit HPV E1 helicase. IFIT3 is shown to enhance TBK1 and MAVS interation to potentiate antiviral response. IFIT5 is reported to facilitate the interaction between I B kinase (IKK) and TGF- -activated kinase 1 (TAK1) by binding both proteins, resulting in enhanced NF- B activation as a reuslt of increased IKK phosphorylation and activation. In addition, both IFIT1 and IFIT5 are shown to recognize viral RNA carrying 5 -triphosphate (PPP-RNA). Human IFIT5 contains eight tetratricopeptide repeat (TPR) helix-turn-helix motifs (a.a. 51-84, 94-127, 138-173, 181-214, 249-282, 338-371, 376-410, 435-468), with a.a. 254-260 within TPR repeat 5 mediating PPP-RNA interaction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p75NTR
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antibody serum with 0.05% Sodium Azide.
Gene Ngfr, Tnfrsf16
Gene Accession No. P07174
Host Species Rabbit
Immunogen GST-tagged recombinant rat p75NTR ICD fragment.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_036742
Test Specificity This antiserum detected the ∽75 kDa full-length p75NTR (neurotrophin receptor) as well as the ∽30 kDa CTF and the ∽25 kDa ICD cleavage fragments by immunoprecipitation and Western blotting (Kenchappa, R.S., et al. (2006). Neuron. 50(2):219-232).
Target Species Chicken
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.