Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ p8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14177
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14177 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14177 Supplier Invitrogen™ Supplier No. PA14177
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4177 detects human p8 (com1). PA1-4177 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~9 kDa protein representing Human p8 (com1) in HepGe cell lysates. The PA1-4177 immunogen is a synthetic peptide corresponding to residues Q(13) P P G P E D E D S S L D E S D(28) of human p8.

The human p8 gene encodes a nuclear protein with strong mitogenic activity, which is expressed in the pancreas in response to acute pancreatitis. Response to TNF alpha and staurosporine-induced apoptosis is not modified by p8 expression. However, cells expressing p8 grow significantly and more rapidly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p8
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene NUPR1
Gene Accession No. O60356
Gene Alias 2310032H04Rik; 60B8AG; CAGA; Candidate of metastasis 1; CFAG; CGLA; COM1; CP-10; L1Ag; MA387; MIF; MRP8; NIF; Nuclear protein 1; nuclear protein 1, transcriptional regulator; nuclear protein 1-like; nuclear protein transcription regulator 1; nuclear protein, transcriptional regulator, 1; nuclear proten 1; nuclear transcriptional regulator protein 1; NUPR1; P8; protein p8
Gene Symbols NUPR1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues Q(13) P P G P E D E D S S L D E S D(28) of human p8.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26471
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.