Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P8 Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502278026
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8026 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8026 Supplier Leinco Technologies Inc Supplier No. 41011100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

P8 Polyclonal antibody specifically detects P8 in Human samples. It is validated for Western Blot

Specifications

Antigen P8
Applications Western Blot
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene NUPR1
Gene Accession No. O60356
Gene Alias 2310032H04Rik; 60B8AG; CAGA; Candidate of metastasis 1; CFAG; CGLA; COM1; CP-10; L1Ag; MA387; MIF; MRP8; NIF; Nuclear protein 1; nuclear protein 1, transcriptional regulator; nuclear protein 1-like; nuclear protein transcription regulator 1; nuclear protein, transcriptional regulator, 1; nuclear proten 1; nuclear transcriptional regulator protein 1; NUPR1; P8; protein p8
Host Species Rabbit
Immunogen Peptide corresponding to the C-terminus of human p8 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26471
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.