Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p90 RSK1 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501992032
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2032 25 μL
50-199-2031 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2032 Supplier Rockland Immunochemicals Supplier No. 600401C66S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for human p90 RSK1 protein. A BLAST analysis was used to suggest cross reactivity with p90 RSK1 from human, rat, mouse, horse, bovine, and dog based on 100% homology with the immunizing sequence. Cross reactivity with p90 RSK1 pS732 from other sources has not been determined.

This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

Specifications

Antigen p90 RSK1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene RPS6KA1
Gene Accession No. P18653, Q15418, Q63531
Gene Alias 90 kDa ribosomal protein S6 kinase 1; CLS; dJ590P13.1 (ribosomal protein S6 kinase, 90kD, polypeptide 1); HU-1; HU-2; HU-3; ISPK-1; MAP kinase-activated protein kinase 1; MAP kinase-activated protein kinase 1a; MAPK-activated protein kinase 1; MAPK-activated protein kinase 1a; MAPKAP kinase 1; MAPKAP kinase 1a; MAPKAP kinase 1c; MAPKAPK-1; Mapkapk1a; Mapkapk-1a; MAPKAPK1B; MAPKAPK1C; MAPKAPK-1c; MRX19; OTTHUMP00000004114; OTTMUSP00000010160; p90Rsk; p90-RSK; p90-RSK 1; p90-RSK 2; p90Rsk1; p90-Rsk1; p90RSK2; p90-RSK2; p90-RSK3; p90S6K; pp90RSK1; pp90RSK2; pp90RSK3; Ribosomal protein S6 kinase 2 alpha; ribosomal protein S6 kinase 2 beta; ribosomal protein S6 kinase a, polypeptide 1; ribosomal protein S6 kinase A1; ribosomal protein S6 kinase A1 L homeolog; ribosomal protein S6 kinase alpha 1; Ribosomal protein S6 kinase alpha-1; Ribosomal protein S6 kinase II alpha; ribosomal protein S6 kinase II beta; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase, 90kD, polypept
Gene Symbols RPS6KA1
Host Species Rabbit
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100057166, 20111, 487353, 533908, 6195, 81771
Target Species Bovine, Canine, Equine, Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.