Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p97/VCP Antibody (5H12), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | p97/VCP |
---|---|
Clone | 5H12 |
Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry |
Classification | Monoclonal |
Description
p97/VCP Monoclonal antibody specifically detects p97/VCP in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, ImmunocytochemistrySpecifications
p97/VCP | |
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
15S Mg(2+)-ATPase p97 subunit, IBMPFDMGC8560, MGC131997, p97, TER ATPase, TERA, transitional endoplasmic reticulum ATPase, valosin containing protein, Valosin-containing proteinMGC148092, yeast Cdc48p homolog | |
A synthesized peptide derived from Human p97/VCP [UniProt P55072] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
5H12 | |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Cellular Markers, DNA Repair, Membrane Trafficking and Chaperones, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
7415 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title