Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p97/VCP Antibody (Hs-14), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB500455
Description
p97/VCP Monoclonal specifically detects p97/VCP in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
p97/VCP | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:1000, Flow Cytometry 3 to 12 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL | |
VCP | |
Freshly ejaculated human sperms were washed in PBS and extracted in 3% acetic acid, 10% glycerol, 30 mM benzaminidine. The acid extract was dialyzed against 0.2% acetic acid and subsequently used for immunization. | |
Affinity Purified | |
RUO | |
7415 | |
Human, Mouse | |
IgM |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
Hs-14 | |
Unconjugated | |
15S Mg(2+)-ATPase p97 subunit, IBMPFDMGC8560, MGC131997, p97, TER ATPase, TERA, transitional endoplasmic reticulum ATPase, valosin containing protein, Valosin-containing proteinMGC148092, yeast Cdc48p homolog | |
Mouse | |
220 kDa | |
0.1 mg | |
Primary | |
The antibody Hs-14 reacts with VCP (valosin-containing protein) a 220 kDa protein previously identified under the general name “intra-acrosomal protein”. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction