Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PA 8
GREENER_CHOICE

Catalog No. 665710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-571-0 10 mg
66-575-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 66-571-0 Supplier Tocris Bioscience™ Supplier No. 6657/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

PAC1 receptor antagonist

PA 8 is a PAC1 receptor antagonist. Inhibits PACAP-induced CREB phosphorylation in PAC1, but not VPAC1 or VPAC2, expressing cells. Also inhibits PACAP-induced increases in cAMP levels in vitro (IC50 = 2 nM) and PACAP-induced aversive responses and mechanical allodynia following intrathecal injection in vivo.

Specifications

Chemical Name or Material 2-Amino-5,8-dihydro-5-[3-methoxy-4-(2-propen-1-yloxy)phenyl]pyrido[2,3-d]pyrimidine-4,7(3H,6H)-dione
CAS 878437-15-1
Quantity 10 mg
Molecular Formula C17H18N4O4
Formula Weight 342.35
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.