Learn More
Description
Application: Live-cell imaging as a self-labeling tag substrate. Suitable for confocal microscopy, super-resolution microscopy (SRM) techniques including PALM (in both live and fixed cells). Allows multicolor single tracking experiments.
Properties and Photophysical Data: PA Janelia Fluor™ 549, Haloalkane shows superior brightness and photostability.
Janelia Fluor™ is a registered trademark of Howard Hughes Medical Institute.
Specifications
Specifications
| Chemical Name or Material | 3',6'-Bis(1-azetidinyl)-N-[2-[2-[(6-chlorohexyl)oxy]ethoxy]ethyl]-2-diazo-2,3-dihydro-3-oxospiro[1H-indene-1,9'-[9H]xanthene]-6-carboxamide |
| CAS | 1811539-43-1 |
| Quantity | 10 μg |
| Product Line | Tocris |
| Target | Photoactivatable Dyes |
| Molecular Formula | C38H42ClN5O5 |
| Product Type | Bioactive Small Molecule |
| Purity | 90% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
