Learn More
Description
Application: Suitable for flow cytometry, confocal microscopy, super resolution microscopy (SRM) including dSTORM (in both live and fixed cells) and STED.
Properties and Photophysical Data: Excitation and emission maxima (λ) are 551 - 553 nm and 570 - 573 nm, respectively.
Please see the product protocol for further information and a guide to protein/antibody labeling.
Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.
Specifications
Specifications
| Chemical Name or Material | 3',6'-Di(azetidin-1-yl)-2-diazo-N-(2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)-3-oxo-2,3-dihydrospiro[indene-1,9'-xanthene]-6-carboxamide |
| Quantity | 300 μg |
| Target | Photoactivatable Dyes |
| Molecular Formula | C34H28N6O5 |
| Purity | 90% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
