Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PA1 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP106541C
Description
PA1 Polyclonal antibody specifically detects PA1 in Human samples. It is validated for Western BlotSpecifications
PA1 | |
Polyclonal | |
Western Blot | |
chromosome 16 open reading frame 53, FLJ22459, GAS, glutamate-rich coactivator interacting with SRC1/NCOA1, MGC4606, PA1, PAXIP1-associated protein 1, PTIP-associated 1 protein, PTIP-associated protein 1 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human PTIP-associated 1 protein using the numbering given in entry NP_078792.1 (GeneID 79447). | |
0.1 mL | |
Signal Transduction | |
79447 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction