Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PA2140 Mouse anti-P. aeruginosa, APC, Clone: 1F5, Abnova™

Catalog No. 89339101 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-101 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-101 Supplier Abnova Corporation Supplier No. MAB19816
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

PA2140 Monoclonal antibody specifically detects PA2140 in P. aeruginosa samples. It is validated for WB, ELISA.

Specifications

Antigen PA2140
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1F5
Conjugate APC
Description putative metallothionein
Gene PA2140
Host Species Mouse
Immunogen Recombinant protein corresponding to full length of Pseudomonas aeruginosa Metallothionein
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 880550
Target Species P. aeruginosa
Content And Storage Store at -20°C
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.