Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PA2140 Mouse anti-P. aeruginosa, HRP, Clone: 1F5, Abnova™
Mouse Monoclonal Antibody.
Supplier: Abnova Corporation MAB19818
Description
PA2140 Monoclonal antibody specifically detects PA2140 in P. aeruginosa samples. It is validated for WB, ELISA.Specifications
PA2140 | |
Monoclonal | |
HRP | |
PA2140 | |
Recombinant protein corresponding to full length of Pseudomonas aeruginosa Metallothionein | |
Primary | |
P. aeruginosa | |
Liquid |
Western Blot, ELISA | |
1F5 | |
putative metallothionein | |
Mouse | |
100 μg | |
880550 | |
Store at -20°C | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction