Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PA28 Activator alpha Subunit/PSME1 Mouse anti-Human, PE, Clone: 12H3, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259434PE
This item is not returnable.
View return policy
Description
PA28 Activator alpha Subunit/PSME1 Monoclonal antibody specifically detects PA28 Activator alpha Subunit/PSME1 in Human samples. It is validated for Flow CytometrySpecifications
PA28 Activator alpha Subunit/PSME1 | |
Monoclonal | |
PE | |
11S regulator complex alpha subunit, 11S regulator complex subunit alpha, 29-kD MCP activator subunit, IFI5111MGC8628, IGUP I-5111, Interferon gamma up-regulated I-5111 protein, interferon-gamma IEF SSP 5111, interferon-gamma-inducible protein 5111, PA28a, PA28alphaActivator of multicatalytic protease subunit 1, proteasome (prosome, macropain) activator subunit 1 (PA28 alpha), Proteasome activator 28 subunit alpha, proteasome activator complex subunit 1, proteasome activator subunit-1, REGalpha, REG-alpha | |
Recombinant human PA28 Activator alpha Subunit/PSME1 (1-249aa) purified from E. coli | |
0.1 mL | |
Cell Biology | |
5720 | |
Store at 4°C in the dark. | |
IgG2a κ |
Flow Cytometry | |
12H3 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction