Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PA28 Activator alpha Subunit/PSME1 Rabbit anti-Human, Clone: 16, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | PA28 Activator alpha Subunit/PSME1 |
---|---|
Clone | 16 |
Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000 |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
PA28 Activator alpha Subunit/PSME1 Monoclonal specifically detects PA28 Activator alpha Subunit/PSME1 in Human samples. It is validated for Western Blot, ELISA.Specifications
PA28 Activator alpha Subunit/PSME1 | |
Western Blot 1:500-1:2000, ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
5720 | |
IgG | |
Protein A purified |
16 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Human | |
11S regulator complex alpha subunit, 11S regulator complex subunit alpha, 29-kD MCP activator subunit, IFI5111MGC8628, IGUP I-5111, Interferon gamma up-regulated I-5111 protein, interferon-gamma IEF SSP 5111, interferon-gamma-inducible protein 5111, PA28a, PA28alphaActivator of multicatalytic protease subunit 1, proteasome (prosome, macropain) activator subunit 1 (PA28 alpha), Proteasome activator 28 subunit alpha, proteasome activator complex subunit 1, proteasome activator subunit-1, REGalpha, REG-alpha | |
Recombinant Human PA28 Activator alpha Subunit/PSME1 Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title