Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PA28 Activator alpha Subunit/PSME1 Rabbit anti-Human, Mouse, Rat, Clone: 6H7X3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | PA28 Activator alpha Subunit/PSME1 |
---|---|
Clone | 6H7X3 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
PA28 Activator alpha Subunit/PSME1 Monoclonal antibody specifically detects PA28 Activator alpha Subunit/PSME1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
PA28 Activator alpha Subunit/PSME1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Biology | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5720 | |
IgG | |
Affinity purified |
6H7X3 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
11S regulator complex alpha subunit, 11S regulator complex subunit alpha, 29-kD MCP activator subunit, IFI5111MGC8628, IGUP I-5111, Interferon gamma up-regulated I-5111 protein, interferon-gamma IEF SSP 5111, interferon-gamma-inducible protein 5111, PA28a, PA28alphaActivator of multicatalytic protease subunit 1, proteasome (prosome, macropain) activator subunit 1 (PA28 alpha), Proteasome activator 28 subunit alpha, proteasome activator complex subunit 1, proteasome activator subunit-1, REGalpha, REG-alpha | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-115 of human PA28 Activator alpha Subunit/PSME1 (Q06323). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title