Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PA28 Activator alpha Subunit/PSME1 Rabbit anti-Human, Mouse, Rat, Clone: JU60-33, Novus Biologicals™
SDP

Catalog No. NBP275606 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275606 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275606 Supplier Novus Biologicals Supplier No. NBP275606

Rabbit Monoclonal Antibody

PA28 Activator alpha Subunit/PSME1 Monoclonal specifically detects PA28 Activator alpha Subunit/PSME1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen PA28 Activator alpha Subunit/PSME1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JU60-33
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias 11S regulator complex alpha subunit, 11S regulator complex subunit alpha, 29-kD MCP activator subunit, IFI5111MGC8628, IGUP I-5111, Interferon gamma up-regulated I-5111 protein, interferon-gamma IEF SSP 5111, interferon-gamma-inducible protein 5111, PA28a, PA28alphaActivator of multicatalytic protease subunit 1, proteasome (prosome, macropain) activator subunit 1 (PA28 alpha), Proteasome activator 28 subunit alpha, proteasome activator complex subunit 1, proteasome activator subunit-1, REGalpha, REG-alpha
Gene Symbols PSME1
Host Species Rabbit
Immunogen Recombinant protein within Human PA28 Activator alpha Subunit/PSME1 aa 1-155/ 249. (SwissProt: Q06323 Human; SwissProt: P97371 Mouse; SwissProt: Q63797 Rat)
Molecular Weight of Antigen 27 kDa
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 5720.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.