Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PA28 Activator beta Subunit/PSME2 Antibody (06), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306553V
Description
PA28 Activator beta Subunit/PSME2 Monoclonal antibody specifically detects PA28 Activator beta Subunit/PSME2 in Human samples. It is validated for Western Blot, ELISASpecifications
PA28 Activator beta Subunit/PSME2 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
06 | |
Western Blot, ELISA | |
11S regulator complex beta subunit, 11S regulator complex subunit beta, Activator of multicatalytic protease subunit 2, MCP activator, 31-kD subunit, PA28b, PA28betacell migration-inducing protein 22, proteasome (prosome, macropain) activator subunit 2 (PA28 beta), Proteasome activator 28 subunit beta, proteasome activator 28-beta, proteasome activator complex subunit 2, proteasome activator hPA28 subunit beta, REGbeta, REG-beta | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PA28 Activator beta Subunit/PSME2 (PSME2_HUMAN; Met1-Tyr239). | |
0.1 mL | |
Neuroscience, Ubiquitin Proteasome Pathway | |
5721 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction